52587
Hexakis(2,3,6-tri-O-octyl)-α-cyclodextrin
purum, ≥97.0% (TLC)
Synonym(s):
2,3,6-Tri-O-octyl-α-cyclodextrin
About This Item
grade
purum
assay
≥97.0% (TLC)
density
1.14 g/mL at 20 °C (lit.)
SMILES string
CCCCCCCCOC[C@H]1O[C@@H]2O[C@@H]3[C@@H](COCCCCCCCC)O[C@H](O[C@@H]4[C@@H](COCCCCCCCC)O[C@H](O[C@@H]5[C@@H](COCCCCCCCC)O[C@H](O[C@@H]6[C@@H](COCCCCCCCC)O[C@H](O[C@@H]7[C@@H](COCCCCCCCC)O[C@H](O[C@H]1[C@H](OCCCCCCCC)[C@H]2OCCCCCCCC)[C@H](OCCCCCCCC)[C@H]7OCCCCCCCC)[C@H](OCCCCCCCC)[C@H]6OCCCCCCCC)[C@H](OCCCCCCCC)[C@H]5OCCCCCCCC)[C@H](OCCCCCCCC)[C@H]4OCCCCCCCC)[C@H](OCCCCCCCC)[C@H]3OCCCCCCCC
InChI
1S/C180H348O30/c1-19-37-55-73-91-109-127-181-145-151-157-163(187-133-115-97-79-61-43-25-7)169(193-139-121-103-85-67-49-31-13)175(199-151)206-158-152(146-182-128-110-92-74-56-38-20-2)201-177(171(195-141-123-105-87-69-51-33-15)164(158)188-134-116-98-80-62-44-26-8)208-160-154(148-184-130-112-94-76-58-40-22-4)203-179(173(197-143-125-107-89-71-53-35-17)166(160)190-136-118-100-82-64-46-28-10)210-162-156(150-186-132-114-96-78-60-42-24-6)204-180(174(198-144-126-108-90-72-54-36-18)168(162)192-138-120-102-84-66-48-30-12)209-161-155(149-185-131-113-95-77-59-41-23-5)202-178(172(196-142-124-106-88-70-52-34-16)167(161)191-137-119-101-83-65-47-29-11)207-159-153(147-183-129-111-93-75-57-39-21-3)200-176(205-157)170(194-140-122-104-86-68-50-32-14)165(159)189-135-117-99-81-63-45-27-9/h151-180H,19-150H2,1-18H3/t151-,152-,153-,154-,155-,156-,157-,158-,159-,160-,161-,162-,163+,164+,165+,166+,167+,168+,169-,170-,171-,172-,173-,174-,175-,176-,177-,178-,179-,180-/m1/s1
InChI key
RMPVMBYXMRFORE-YHIDQUJYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service