Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52206

Supelco

1,7-Hexadecadiene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)4CH=CH(CH2)7CH3
CAS Number:
Molecular Weight:
222.41
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥90% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C\CCCCC=C

InChI

1S/C16H30/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3,13,15H,1,4-12,14,16H2,2H3/b15-13+

InChI key

ZKGUVFWOQHIDON-FYWRMAATSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ki-Chang Lee et al.
Journal of agricultural and food chemistry, 68(9), 2803-2815 (2020-02-11)
This study evaluated the applicability of a rapid analytical method using a headspace solid-phase microextraction gas chromatography/mass spectrometry (HS-SPME-GC/MS) technique to identify gamma-irradiated soybeans (0.1-5 kGy). From the partial least squares discriminant analysis used to discriminate between non-irradiated and irradiated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service