Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52018

Supelco

[(3R)-3-Hydroxy-cis-tetradec-9-enoyl]-L-carnitine

analytical standard

Synonym(s):

(2R)-3-Carboxy-2-[[(3R,9Z)-3-hydroxy-1-oxo-9-tetradecen-1-yl]oxy]-N,N,N-trimethyl-1-propanaminium inner salt, (3R)-3-Hydroxymyristoleoyl-L-carnitine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H39NO5
Molecular Weight:
385.54
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥84.0% (HPLC)

form

solid

optical activity

[α]/D -20.0±2.0°, c = 0.1 in methanol

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

storage temp.

−20°C

SMILES string

C[N+](C)(C)C[C@H](OC(C[C@H](O)CCCCC/C=C\CCCC)=O)CC([O-])=O

InChI key

JPNOKZQZVLXQNC-FAPWYTRCSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service