Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51876

Supelco

Polystrene Standard 88′000

certified reference material (acc. to BAM), certified by BAM, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Polystyrene, PS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]n
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material (acc. to BAM)

agency

certified by the BSC

quality

certified by BAM

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Exact molecular weight Mw, expiry date and detailed informaton certification can be found on the certificate and certification report, included in each package.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service