Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51834

Supelco

Sodium 1-heptanesulfonate solution

suitable for ion pair chromatography, concentrate, LiChropur, ampule

Synonym(s):

1-Heptanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH2SO3Na
CAS Number:
Molecular Weight:
202.25
Beilstein/REAXYS Number:
3731762
MDL number:
UNSPSC Code:
41115711
PubChem Substance ID:

description

anionic

Quality Level

quality

LiChropur
ampule

packaging

ampule of 25 mL

technique(s)

HPLC: suitable
ion pair chromatography: suitable

λ

in concentrate

UV absorption

λ: 210 nm Amax: 0.07
λ: 220 nm Amax: 0.04
λ: 230 nm Amax: 0.03
λ: 260 nm Amax: 0.01
λ: 500 nm Amax: 0.01

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na].CCCCCCCS(O)(=O)=O

InChI

1S/C7H16O3S.Na/c1-2-3-4-5-6-7-11(8,9)10;/h2-7H2,1H3,(H,8,9,10);/q;+1/p-1

InChI key

REFMEZARFCPESH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Concentrate for 6x1 L ready-to-use solution. Dilute contents of one ampule (~0.3 M) to 1 L with HPLC grade water to obtain a 0.005 M eluent solution.

Application

Sodium 1-heptanesulfonate may be used as an ion-pairing reagent for the determination of ethambutol in pharmaceutical dosage forms by ion-pair reversed phase liquid chromatography with UV detection.[1]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of ethambutol by ion-pair reversed phase liquid chromatography with UV detection.
Jiang Z, et al.
Analytica Chimica Acta, 456(2), 189- 192 (2002)
Marianna Sikorska et al.
Neurobiology of aging, 35(10), 2329-2346 (2014-04-30)
Although the support for the use of antioxidants, such as coenzyme Q(10) (CoQ(10)), to treat Parkinson's disease (PD) comes from the extensive scientific evidence, the results of conducted thus far clinical trials are inconclusive. It is assumed that the efficacy
Takeshi Oshizaka et al.
International journal of pharmaceutics, 475(1-2), 292-297 (2014-08-27)
Skin concentrations of topically administered compounds need to be considered in order to evaluate their efficacies and toxicities. This study investigated the relationship between the skin permeation and concentrations of compounds, and also predicted the skin concentrations of these compounds
Miriam Dadparvar et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 88(2), 510-517 (2014-07-06)
Severe intoxications with organophosphates require the immediate administration of atropine in combination with acetyl cholinesterase (AChE) reactivators such as HI-6. Although this therapy regimen enables the treatment of peripheral symptoms, the blood-brain barrier (BBB) restricts the access of the hydrophilic
Jie Ma et al.
Pharmacology, 93(5-6), 220-224 (2014-07-12)
It is significant for patients with traumatic brain injury (TBI) to receive prehospital emergency treatment as early as possible to reduce mortality. Therefore, a new prefilled multi-drug injector was developed to improve the treatment of TBI. Here, we studied the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service