51754
Carbamazepine-2,3,4,6,7,8,10,11-d8
analytical standard
Synonym(s):
5H-Dibenz[b,f]azepine-2,3,4,6,7,8,10,11-d8-5-carboxamide, [2H8]-Carbamazepine
About This Item
grade
analytical standard
Quality Level
assay
≥98.5% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
storage temp.
2-8°C
SMILES string
[2H]C1=C([2H])C(C=C([2H])C([2H])=C2[2H])=C2N(C(N)=O)C3=C1C=C([2H])C([2H])=C3[2H]
InChI
1S/C15H12N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H,(H2,16,18)/i1D,2D,3D,4D,7D,8D,9D,10D
InChI key
FFGPTBGBLSHEPO-PKSNNKEVSA-N
Application
Analysis Note
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1A - STOT SE 3
target_organs
Central nervous system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service