About This Item
83 mmHg ( 37.7 °C)
Recommended Products
vapor density
3.5 (vs air)
Quality Level
vapor pressure
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)
grade
puriss. p.a.
assay
≥99.5% (GC)
form
liquid
autoignition temp.
433 °F
expl. lim.
7 %
technique(s)
GC/GC: suitable
impurities
≤0.05% water
evapn. residue
≤0.002%
refractive index
n20/D 1.387 (lit.)
n20/D 1.388
bp
98 °C (lit.)
mp
−91 °C (lit.)
density
0.684 g/mL at 20 °C
0.684 g/mL at 25 °C (lit.)
cation traces
Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg
SMILES string
CCCCCCC
InChI
1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3
InChI key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Other Notes
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
24.8 °F - closed cup
flash_point_c
-4 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service