Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51505

Supelco

Metronidazole-(ethylene-d4) hydrochloride

analytical standard

Synonym(s):

[2H4]-Metronidazol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4H5N3O3 · HCl
CAS Number:
Molecular Weight:
211.64
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[2H]C(C([2H])([2H])O)([2H])N1C(C)=NC=C1[N+]([O-])=O.Cl

InChI

1S/C6H9N3O3.ClH/c1-5-7-4-6(9(11)12)8(5)2-3-10;/h4,10H,2-3H2,1H3;1H/i2D2,3D2;

InChI key

FPTPAIQTXYFGJC-DAHDXRBSSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

LC-MS identity conforms

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinny Jeffery et al.
Clinical biochemistry, 50(6), 323-330 (2016-12-10)
Metronidazole is an oral antibiotic which is widely used in the treatment of patients with Clostridium difficile associated disease. This article describes the validation of a LC-MS/MS assay for the measurement of metronidazole in human faecal samples. Matrix matched and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service