Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51479

Supelco

Leaded brass

manufactured by MBH Analytical Ltd (31X 7835.3)

Synonym(s):

Copper-zinc-lead alloy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

technique(s)

XRF: suitable

format

matrix material

Analysis Note

Values of analytes vary lot to lot.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service