Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51386

Supelco

1,4-Benzoquinone

pharmaceutical secondary standard; traceable to USP

Synonym(s):

p-Benzoquinone, Quinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(=O)2
CAS Number:
Molecular Weight:
108.09
Beilstein/REAXYS Number:
773967
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

certified reference material
pharmaceutical secondary standard

agency

traceable to USP 1056504

vapor density

3.73 (vs air)

vapor pressure

0.1 mmHg ( 25 °C)

autoignition temp.

815 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

113-115 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

O=C1C=CC(=O)C=C1

InChI

1S/C6H4O2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

AZQWKYJCGOJGHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Vandana Bhalla et al.
Dalton transactions (Cambridge, England : 2003), 42(13), 4464-4469 (2013-02-16)
The binding behavior of nanoaggregates of pentacenequinone derivative 1 having thiophene groups has been investigated toward various cations (Fe(3+), Fe(2+), Pd(2+), Mg(2+), Cd(2+), Hg(2+), Ni(2+), Zn(2+), Cu(2+), Pb(2+), Co(2+), K(+), Na(+) and Li(+)) using UV-vis and fluorescence spectroscopy. Among the
S Saif Hasan et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(11), 4297-4302 (2013-02-27)
As much as two-thirds of the proton gradient used for transmembrane free energy storage in oxygenic photosynthesis is generated by the cytochrome b6f complex. The proton uptake pathway from the electrochemically negative (n) aqueous phase to the n-side quinone binding
K He et al.
Biochemical pharmacology, 62(2), 191-198 (2001-06-08)
Oxidative ring opening of troglitazone (TGZ)(1) a thiazolidine 2,4-dione derivative used for the treatment of type II diabetes mellitus, leads to the formation of a quinone metabolite. The formation of TGZ quinone was shown to be NADPH dependent and to
K Roberg et al.
Free radical biology & medicine, 27(11-12), 1228-1237 (2000-01-21)
Apoptosis was induced in human foreskin fibroblasts by the redox-cycling quinone naphthazarin (5,8-dihydroxy-1,4-naphthoquinone). Most of the cells displayed ultrastructure typical of apoptosis after 8 h of exposure to naphthazarin. Apoptosis was inhibited in fibroblasts pretreated with the cathepsin D inhibitor
Jens Tiefenbach et al.
Disease models & mechanisms, 11(9) (2018-09-02)
Current peroxisome proliferator-activated receptor (PPAR)-targeted drugs, such as the PPARγ-directed diabetes drug rosiglitazone, are associated with undesirable side effects due to robust agonist activity in non-target tissues. To find new PPAR ligands with fewer toxic effects, we generated transgenic zebrafish

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service