Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50956

Supelco

Stevioside

analytical standard

Synonym(s):

(4α)-13-[(2-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid β-D-glucopyranosyl ester, Steviosin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H60O18
CAS Number:
Molecular Weight:
804.87
EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water (Karl Fischer)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)[C@](CC[C@@H]23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(=O)O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O

InChI

1S/C38H60O18/c1-16-11-37-9-5-20-35(2,7-4-8-36(20,3)34(50)55-32-29(49)26(46)23(43)18(13-40)52-32)21(37)6-10-38(16,15-37)56-33-30(27(47)24(44)19(14-41)53-33)54-31-28(48)25(45)22(42)17(12-39)51-31/h17-33,39-49H,1,4-15H2,2-3H3/t17-,18-,19-,20+,21+,22-,23-,24-,25+,26+,27+,28-,29-,30-,31+,32+,33+,35-,36-,37-,38+/m1/s1

InChI key

UEDUENGHJMELGK-HYDKPPNVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Component of stevia.
Stevioside is a diterpene glycoside, naturally extracted from the leaves of Stevia rebaudiana Bertoni plant[1], which can find wide applications as a commercial non-caloric sweetener and can also be used in the treatment of diabetes.[2]
[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Stevioside
Guens C.MJ
Phytochemistry, 64, 913-921 (2003)
Efficacy and tolerability of oral stevioside in patients with mild essential hypertension: a two-year, randomized, placebo-controlled study
Hsieh HM, et al.
Clinical Therapeutics, 25, 2797-2808 (2003)
Munish Puri et al.
Biotechnology advances, 29(6), 781-791 (2011-07-06)
Stevioside is a natural sweetener extracted from leaves of Stevia rebaudiana Bertoni, which is commercially produced by conventional (chemical/physical) processes. This article gives an overview of the stevioside structure, various analysis technique, new technologies required and the advances achieved in
Kumiko Takahashi et al.
Toxicology mechanisms and methods, 22(4), 243-249 (2012-02-10)
In the laboratory, using a PC12 cell system, studies have been conducted on the effects of various chemicals on apoptosis, as it is considered to be an essential part of normal development, maintenance, and defense in organisms. Stevioside is a
Tipwadee Bunprajun et al.
Journal of agricultural and food chemistry, 60(11), 2844-2851 (2012-02-10)
Stevioside, a noncaloric sweetener isolated from Stevia rebaudiana, exhibits anti-inflammatory and immunomodulatory effects through interference of nuclear factor (NF)-kappa B pathway. We investigated whether this anti-inflammatory property of stevioside could improve muscle regeneration following cardiotoxin-induced muscle injury. Adult male Wistar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service