Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50921

Supelco

Methyl 2-nonynoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5C≡CCO2CH3
CAS Number:
Molecular Weight:
168.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard
reference material

assay

≥98.0% (qNMR)
≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

refractive index

n20/D 1.446-1.450
n20/D 1.448 (lit.)

bp

121 °C/20 mmHg (lit.)

density

0.915 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCC#CC(=O)OC

InChI

1S/C10H16O2/c1-3-4-5-6-7-8-9-10(11)12-2/h3-7H2,1-2H3

InChI key

NTLJTUMJJWVCTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

213.8 °F - closed cup

flash_point_c

101 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service