50878
Procyanidin C1
analytical standard
Synonym(s):
Cinnamtannin A1, Proanthocyanidin C1, Procyanidol C1
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥90.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤15.0% water
application(s)
food and beverages
format
neat
shipped in
wet ice
storage temp.
−20°C
SMILES string
O[C@@H]1Cc2c(O)cc(O)c([C@@H]3[C@@H](O)[C@H](Oc4c([C@@H]5[C@@H](O)[C@H](Oc6cc(O)cc(O)c56)c7ccc(O)c(O)c7)c(O)cc(O)c34)c8ccc(O)c(O)c8)c2O[C@@H]1c9ccc(O)c(O)c9
InChI
1S/C45H38O18/c46-18-10-27(54)33-32(11-18)61-42(16-2-5-21(48)25(52)8-16)39(59)37(33)35-29(56)14-30(57)36-38(40(60)43(63-45(35)36)17-3-6-22(49)26(53)9-17)34-28(55)13-23(50)19-12-31(58)41(62-44(19)34)15-1-4-20(47)24(51)7-15/h1-11,13-14,31,37-43,46-60H,12H2/t31-,37-,38+,39-,40-,41-,42-,43-/m1/s1
InChI key
MOJZMWJRUKIQGL-XILRTYJMSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Cocoa-based products by ultra-high performance liquid chromatography coupled to quadrupole Orbitrap high resolution mass spectrometry (UHPLC-Q-Orbitrap-MS/MS).[2]
- Wild apple by UHPLC equipped with diode array detection (DAD) and high resolution multistage mass spectrometry (HRMS).[3]
- Food materials and biological samples by HPLC combined with fluorescence detector (FLD) as well as DAD.[1]
- Lentil seed coats by HPLC-MS with ultraviolet (UV) detection.[4]
- Grape seeds by high-speed counter-current chromatography (HSCCC).[5]
Packaging
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service