50728
Acetochlor-(2-ethyl-6-methylphenyl-d11)
PESTANAL®, analytical standard
Synonym(s):
Acetochlor-d11
About This Item
Recommended Products
grade
analytical standard
Quality Level
isotopic purity
≥98 atom % D
product line
PESTANAL®
assay
≥97.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
O=C(CCl)N(COCC)C1=C(C([2H])([2H])C([2H])([2H])[2H])C([2H])=C([2H])C([2H])=C1C([2H])([2H])[2H]
InChI
1S/C14H20ClNO2/c1-4-12-8-6-7-11(3)14(12)16(10-18-5-2)13(17)9-15/h6-8H,4-5,9-10H2,1-3H3/i1D3,3D3,4D2,6D,7D,8D
InChI key
VTNQPKFIQCLBDU-RLYLBJGXSA-N
Application
Legal Information
signalword
Warning
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service