Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50057

Sigma-Aldrich

Glycerol phosphate calcium salt hydrate

tested according to Ph. Eur.

Synonym(s):

Calcii glycerophosphas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7CaO6P · xH2O
CAS Number:
Molecular Weight:
210.14 (anhydrous basis)
Beilstein/REAXYS Number:
6099192
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

agency

USP/NF
tested according to Ph. Eur.

Quality Level

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

O.[Ca++].OCC(O)COP([O-])([O-])=O

InChI

1S/C3H9O6P.Ca.H2O/c4-1-3(5)2-9-10(6,7)8;;/h3-5H,1-2H2,(H2,6,7,8);;1H2/q;+2;/p-2

InChI key

ILSFDSREGJGPRB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grégory Chatelain et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 20(3), 497-507 (2014-12-24)
Bone is the main target organ for the storage of several toxic metals, including uranium. But the mode of action of uranium on bones remains poorly understood. To better assess the impact of uranium on bone cells, synthetic biomimetic apatites

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service