Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50052

Sigma-Aldrich

Glycine

purum, ≥98.5% (NT)

Synonym(s):

Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein/REAXYS Number:
635782
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:

grade

purum

assay

≥98.5% (NT)

form

powder or crystals

ign. residue

≤0.1% (as SO4)

loss

≤0.2% loss on drying, 110 °C

color

white

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Carvajal-González et al.
Brain : a journal of neurology, 137(Pt 8), 2178-2192 (2014-06-22)
The clinical associations of glycine receptor antibodies have not yet been described fully. We identified prospectively 52 antibody-positive patients and collated their clinical features, investigations and immunotherapy responses. Serum glycine receptor antibody endpoint titres ranged from 1:20 to 1:60 000.
Zoé Husson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(28), 9418-9431 (2014-07-11)
The principal neurons of the cerebellar nuclei (CN), the sole output of the olivo-cerebellar system, receive a massive inhibitory input from Purkinje cells (PCs) of the cerebellar cortex. Morphological evidence suggests that CN principal cells are also contacted by inhibitory
Steven Tran et al.
Behavioural brain research, 276, 161-170 (2014-03-07)
The zebrafish is emerging as a popular animal model for alcohol (ethanol or EtOH) addiction due to its simplicity and practical advantages. Two phenomena associated with ethanol addiction are the development of tolerance and withdrawal. Using a multi-level approach in
Li-Chun Tu et al.
The EMBO journal, 32(24), 3220-3230 (2013-11-12)
Nuclear pore complexes (NPCs) mediate cargo traffic between the nucleus and the cytoplasm of eukaryotic cells. Nuclear transport receptors (NTRs) carry cargos through NPCs by transiently binding to phenylalanine-glycine (FG) repeats on intrinsically disordered polypeptides decorating the NPCs. Major impediments
Suzanne L Grant et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 844(2), 278-282 (2006-08-08)
A simple and versatile methodology using high-performance liquid chromatography (HPLC) with fluorimetric detection was developed to simultaneously determine d-serine along with other metabolically related neuroactive amino acids in the glutamatergic system: L-serine, L-glutamate, L-glutamine, and glycine. On-column sensitivity was in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service