Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50035

Sigma-Aldrich

sn-Glycero-3-phosphocholine 1:1 cadmium chloride adduct

Synonym(s):

L-α-Glycerophosphorylcholine 1:1 cadmium chloride adduct from egg yolk

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H20NO6P · CdCl2
CAS Number:
Molecular Weight:
440.54
Beilstein/REAXYS Number:
4936450
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

animal

form

solid

impurities

≤11% water

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

2-8°C

SMILES string

Cl[Cd]Cl.C[N+](C)(C)CCOP([O-])(=O)OC[C@H](O)CO

InChI

1S/C8H20NO6P.Cd.2ClH/c1-9(2,3)4-5-14-16(12,13)15-7-8(11)6-10;;;/h8,10-11H,4-7H2,1-3H3;;2*1H/q;+2;;/p-2/t8-;;;/m1.../s1

InChI key

RCOWLNSSDOITMP-VFIYQRISSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Starting material for the synthesis of lecithins by diacylation[1]

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

L.D. Bergelson, ed.
Lipid Biochem. Prep. null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service