Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49974

Supelco

(2R)-3-(1-Naphthalenyloxy)-1,2-propanediol

analytical standard

Synonym(s):

(R)-3-(1-Naphthyloxy)-1,2-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14O3
CAS Number:
Molecular Weight:
218.25
Beilstein/REAXYS Number:
3590585
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (HPLC)

optical activity

[α]/D -6.5±1.0°, c = 0.05 in methanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

storage temp.

−20°C

SMILES string

OC[C@@H](O)COC1=CC=CC2=CC=CC=C21

InChI

1S/C13H14O3/c14-8-11(15)9-16-13-7-3-5-10-4-1-2-6-12(10)13/h1-7,11,14-15H,8-9H2/t11-/m1/s1

InChI key

BYNNMWGWFIGTIC-LLVKDONJSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service