Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49949

Supelco

Homosalate-(benzoic ring-d4)

analytical standard

Synonym(s):

2-Hydroxybenzoic-d4 acid 3,3,5-trimethylcyclohexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D4H18O3
Molecular Weight:
266.37
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥98 atom % D

assay

≥98.0% (mixture of isomers, GC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service