Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49903

Sigma-Aldrich

Octyl-β-D-1-thioglucoside 90 mM solution

Synonym(s):

Octyl-β-D-1-thioglucopyranoside solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12161900

storage temp.

2-8°C

InChI

1S/C14H28O5S/c1-2-3-4-5-6-7-8-20-14-13(18)12(17)11(16)10(9-15)19-14/h10-18H,2-9H2,1H3

InChI key

CGVLVOOFCGWBCS-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kraingkrai Ponhong et al.
Talanta, 87, 113-117 (2011-11-22)
A novel simultaneous injection effective mixing analysis system (SIEMA) for determination of direct bilirubin in urine sample was developed. Bilirubin reacts with diazotized sulfanilic acid in the presence of n-octyl-β-d-thioglucoside (OTG) as a solubilizing agent to form OTG-azobilirubin. The flow
H A Chu et al.
Biochemistry, 39(47), 14371-14376 (2000-11-23)
We have developed conditions for recording the low-frequency S(2)/S(1) Fourier transform infrared difference spectrum of hydrated PSII samples. By exchanging PSII samples with buffered (18)O water, we found that a positive band at 606 cm(-)(1) in the S(2)/S(1) spectrum in
Yann Gohon et al.
Biophysical journal, 94(9), 3523-3537 (2008-01-15)
The membrane protein bacteriorhodopsin (BR) can be kept soluble in its native state for months in the absence of detergent by amphipol (APol) A8-35, an amphiphilic polymer. After an actinic flash, A8-35-complexed BR undergoes a complete photocycle, with kinetics intermediate
Kazuo Takeda et al.
Life sciences, 76(19), 2203-2209 (2005-03-01)
This study examined Fe(II)-dependent ATPase activity in OTG (octylthioglucoside) -treated microsomes isolated from Wistar and LEC rats. The ATPase activity of the liver OTG-microsomes from Wistar rats increased sharply in the 5-150 microM range of Fe(II) with a K0.5 value
K Suzuki et al.
The Journal of pharmacology and experimental therapeutics, 288(1), 57-64 (1998-12-23)
A specific sugar-modified peptide has previously been shown to have renal targeting potential in vivo and to have a specific binding site which has been identified in the kidney membrane fraction. In this report, we studied the inhibitory effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service