Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

49892

Supelco

Zinc-aluminum-copper alloy

manufactured by MBH Analytical Ltd (43X Z2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

material

disks

grade

analytical standard

technique(s)

XRF: suitable

diam.

50 mm (20 mm) , dimensions are approximate

format

matrix material

Analysis Note

Values of analytes vary lot to lot.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service