Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

49578

Supelco

4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanone

analytical standard

Synonym(s):

4-(Methyl-d3-nitrosoamino)-1-(3-pyridinyl)-1-butanone, NNK-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D3H10N3O2
CAS Number:
Molecular Weight:
210.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

suitability

corresponds for LC-MS

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+3

Storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])N(CCCC(=O)c1cccnc1)N=O

InChI

1S/C10H13N3O2/c1-13(12-15)7-3-5-10(14)9-4-2-6-11-8-9/h2,4,6,8H,3,5,7H2,1H3/i1D3

InChI key

FLAQQSHRLBFIEZ-FIBGUPNXSA-N

General description

4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanone is a TSNA analytical standard of high-quality for reliable and quantitative analysis of tobacco specific nitrosamines (TSNAs).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsushi Ishizaki et al.
Analytica chimica acta, 1075, 98-105 (2019-06-15)
A simple and sensitive method, using automated online in-tube solid-phase microextraction (SPME) coupled with liquid chromatography-tandem mass spectrometry (LC-MS/MS), was developed for the determination of four tobacco-specific nitrosamines (TSNAs) in mainstream and sidestream smoke of combustion cigarettes and heated tobacco

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service