49498
Quinoxaline
analytical standard
Synonym(s):
1,4-Benzodiazine, Benzo[a]pyrazine, Benzopyrazine
About This Item
grade
analytical standard
Quality Level
assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
bp
220-223 °C (lit.)
mp
29-32 °C (lit.)
density
1.124 g/mL at 25 °C (lit.)
application(s)
cleaning products
clinical
cosmetics
environmental
food and beverages
personal care
format
neat
storage temp.
2-8°C
SMILES string
c1ccc2nccnc2c1
InChI
1S/C8H6N2/c1-2-4-8-7(3-1)9-5-6-10-8/h1-6H
InChI key
XSCHRSMBECNVNS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service