Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

48940

Gitoxigenin

≥98.0% (HPLC)

Synonym(s):

16β-Hydroxydigitoxigenin, 3β,14,16β,21-Tetrahydroxy-20(22)-norcholenic acid lactone, 3β,14,16β-Trihydroxy-5β,20(22)-cardenolide, 5β,20(22)-Cardenolide-3β,14,16β-triol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
UNSPSC Code:
12352200
EC Number:
208-886-2
Beilstein/REAXYS Number:
96483
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

[H][C@]12CC[C@]3([H])[C@]([H])(CC[C@]4(C)[C@H]([C@@H](O)C[C@]34O)C5=CC(=O)OC5)[C@@]1(C)CC[C@H](O)C2

assay

≥98.0% (HPLC)

optical activity

[α]20/D +30±2°, c = 1% in methanol

mp

~230 °C

Biochem/physiol Actions

Structure-activity relationships for 16β-substituted digitoxigenins as inhibitors of Na+/K+ ATPases.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service