Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48933

Sigma-Aldrich

tert-Butyl hydroperoxide solution

~5.5 M in decane

Synonym(s):

1,1-Dimethylethyl hydroperoxide, 2-Hydroperoxy-2-methylpropane, TBHP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COOH
CAS Number:
Molecular Weight:
90.12
Beilstein/REAXYS Number:
1098280
MDL number:
UNSPSC Code:
12352120
PubChem Substance ID:
Pricing and availability is not currently available.

reaction suitability

reagent type: oxidant

concentration

~5.5 M in decane

SMILES string

CC(C)(C)OO

InChI

1S/C4H10O2/c1-4(2,3)6-5/h5H,1-3H3

InChI key

CIHOLLKRGTVIJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Org. Perox. D - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3

flash_point_f

109.4 °F - closed cup

flash_point_c

43 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Łukasz John et al.
Materials science & engineering. C, Materials for biological applications, 78, 901-911 (2017-06-04)
In this paper, we report on synthesis and characterization of three-dimensional biocomposite based on a polymerized 3-(trimethoxysilyl)propyl methacrylate/ethylene glycol dimethacrylate (pTMSPMA/pEGDMA) framework. The resulting composite was doped with Ca
Qiong Tang et al.
Ultrasonics sonochemistry, 20(5), 1168-1175 (2013-03-30)
This work investigated the ultrasonic assisted oxidative desulfurization of bunker-C oil with TBHP/MoO3 system. The operational parameters for the desulfurization procedure such as ultrasonic irradiation time, ultrasonic wave amplitude, catalyst initial concentration and oxidation agent initial concentration were studied. The
Maxim O Ratnikov et al.
Journal of the American Chemical Society, 135(4), 1549-1557 (2013-01-10)
A general mechanism is proposed for transition metal-catalyzed oxidative Mannich reactions of N,N-dialkylanilines with tert-butyl hydroperoxide (TBHP) as the oxidant. The mechanism consists of a rate-determining single electron transfer (SET) that is uniform from 4-methoxy- to 4-cyano-N,N-dimethylanilines. The tert-butylperoxy radical
Yury Minko et al.
Nature protocols, 8(4), 749-754 (2013-03-23)
This protocol describes a new approach for the preparation of stereodefined trisubstituted chiral enolate species, avoiding conventional asymmetric enolization of carbonyl compounds. This protocol was developed as a single-flask synthetic sequence and therefore does not require isolation or purification of
Qicai Xue et al.
Chemical communications (Cambridge, England), 49(35), 3700-3702 (2013-03-29)
A new synthetic approach toward direct C-N bond formation through sp(3) C-H activation has been developed under metal-free conditions. Both primary and secondary benzylic C-H substrates could react smoothly with various amines to give only mono-amination products with good to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service