Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48057

Sigma-Aldrich

Dimethyl 3,4-furandicarboxylate

purum, ≥96.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O5
CAS Number:
Molecular Weight:
184.15
Beilstein/REAXYS Number:
152468
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥96.0% (GC)

bp

91-93 °C/1 mmHg (lit.)

mp

46-49 °C (lit.)
46-49 °C

SMILES string

COC(=O)c1cocc1C(=O)OC

InChI

1S/C8H8O5/c1-11-7(9)5-3-13-4-6(5)8(10)12-2/h3-4H,1-2H3

InChI key

SEVRCIAIDGNMFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service