Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48003

Sigma-Aldrich

3-Furoic acid

purum, ≥98.0% (T)

Synonym(s):

Furan-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O3
CAS Number:
Molecular Weight:
112.08
Beilstein/REAXYS Number:
108638
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (T)

mp

120-122 °C (lit.)
120-122 °C

SMILES string

OC(=O)c1ccoc1

InChI

1S/C5H4O3/c6-5(7)4-1-2-8-3-4/h1-3H,(H,6,7)

InChI key

IHCCAYCGZOLTEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H M Liebich et al.
Journal of chromatography, 525(1), 1-14 (1990-01-26)
Altogether 143 of the organic acids regularly occurring in urine of healthy individuals are identified as methyl esters by gas chromatography-mass spectrometry with respect to their complete chemical structures. They are classified as dicarboxylic acids, oxocarboxylic acids, hydroxycarboxylic acids, aromatic
M J Nozal et al.
Journal of chromatography. A, 917(1-2), 95-103 (2001-06-14)
A high-performance liquid chromatographic method for determining 5-hydroxymethyl-2-furaldehyde (hydroxymethylfurfural), 2-furaldehyde (furfural), furan-2-carboxylic acid (2-furoic acid), furan-3-carboxylic acid (3-furoic acid), furan-3-carboxaldehyde (3-furaldehyde) and 2-aminobenzoic acid methyl ester (methyl anthranilate) in honey and honeydew samples is described. To prevent matrix interference and
Sorin Tunaru et al.
Nature medicine, 9(3), 352-355 (2003-02-04)
Nicotinic acid (niacin), a vitamin of the B complex, has been used for almost 50 years as a lipid-lowering drug. The pharmacological effect of nicotinic acid requires doses that are much higher than those provided by a normal diet. Its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service