Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47287

Sigma-Aldrich

(4-Fluorophenyl)acetone

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2COCH3
CAS Number:
Molecular Weight:
152.17
Beilstein/REAXYS Number:
1936697
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (GC)

refractive index

n20/D 1.496 (lit.)
n20/D 1.496

bp

106-107 °C/18 mmHg (lit.)

density

1.139 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Cc1ccc(F)cc1

InChI

1S/C9H9FO/c1-7(11)6-8-2-4-9(10)5-3-8/h2-5H,6H2,1H3

InChI key

ZUEKIIWSVFBTCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

195.8 °F - closed cup

flash_point_c

91 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service