Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
FC6H4OH
CAS Number:
Molecular Weight:
112.10
EC Number:
206-748-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1904536
MDL number:
grade
purum
assay
≥97.0% (GC)
refractive index
n20/D 1.514 (lit.), n20/D 1.516
bp
178 °C (lit.)
mp
8-12 °C (lit.)
density
1.238 g/mL at 25 °C (lit.)
SMILES string
Oc1cccc(F)c1
InChI
1S/C6H5FO/c7-5-2-1-3-6(8)4-5/h1-4,8H
InChI key
SJTBRFHBXDZMPS-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.