Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47148

Supelco

Amidoflumet

analytical standard

Synonym(s):

5-Chloro-2-{[(trifluoromethyl)sulfonyl]amino}benzoic acid methyl ester, Methyl 5-chloro-2-{[(trifluoromethyl)sulfonyl]amino}benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7ClF3NO4S
CAS Number:
Molecular Weight:
317.67
UNSPSC Code:
77101502

grade

analytical standard

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CS(=O)(=O)Nc1ccc(Cl)cc1C(=O)OC(F)(F)F

InChI

1S/C9H7ClF3NO4S/c1-18-8(15)6-4-5(10)2-3-7(6)14-19(16,17)9(11,12)13/h2-4,14H,1H3

Inchi Key

KBHDSWIXRODKSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Miticide for house dust mites

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service