Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46901

Supelco

N4-Phthalylsulfathiazole

VETRANAL®, analytical standard

Synonym(s):

2-[[[4-[(2-Thiazolylamino)sulfonyl]phenylamino]carbonyl]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13N3O5S2
CAS Number:
Molecular Weight:
403.43
Beilstein/REAXYS Number:
359901
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

240 °C (dec.) (lit.)

application(s)

clinical testing

format

neat

SMILES string

OC(=O)c1ccccc1C(=O)Nc2ccc(cc2)S(=O)(=O)Nc3nccs3

InChI

1S/C17H13N3O5S2/c21-15(13-3-1-2-4-14(13)16(22)23)19-11-5-7-12(8-6-11)27(24,25)20-17-18-9-10-26-17/h1-10H,(H,18,20)(H,19,21)(H,22,23)

InChI key

PBMSWVPMRUJMPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>734.0 °F

flash_point_c

> 390 °C

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

INTESTINAL ANTISEPSIS.
I COHN
Diseases of the colon and rectum, 8, 11-18 (1965-01-01)
A C Turner
Scandinavian journal of gastroenterology. Supplement, 84, 106-110 (1983-01-01)
The chemoprophylaxis of travellers' diarrhoea by iodochlorhydroxyquinoline combinations, neomycin and phthalsulphathiazole, furazolidone, bismuth salicylates and streptotriad is reviewed. Streptotriad is regarded as a highly effective prophylactic against travellers' diarrhoea. Some elementary food hygiene precautions are detailed.
Studies on porcine enteritis. II. Experimental therapy with sulfathalidine, sulfamethazine, sodium arsanilate, and bacitracin in a natural outbreak of swine dysentery.
L E BOLEY et al.
The Cornell veterinarian, 41(3), 231-235 (1951-07-01)
The effects of bowel preparation for colon surgery on the colon microflora.
G P Gerritsen et al.
The Netherlands journal of surgery, 34(2), 67-71 (1982-05-01)
Modification of the acute radiation syndrome. I Alteration by neomycin-sulfathalidine therapy.
E J POTH et al.
Texas reports on biology and medicine, 9(3), 645-651 (1951-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service