Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46657

Supelco

Prednisolone 21-acetate

VETRANAL®, analytical standard

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-3,20-dione 21-acetate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate, 21-Acetoxy-1,4-pregnadiene-11β,17α-diol-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30O6
CAS Number:
Molecular Weight:
402.48
Beilstein/REAXYS Number:
3111798
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-18,20,26,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1

InChI key

LRJOMUJRLNCICJ-JZYPGELDSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael Campagna et al.
European radiology, 23(2), 424-427 (2012-08-07)
To evaluate the occurrence of coring after needle insertion through the rubber stopper of prednisolone acetate vials. Two-hundred vials of prednisolone acetate were randomly distributed to two radiologists. Prednisolone acetate was drawn up through the rubber bung of the vials
Nancy J McLean et al.
Journal of the American Veterinary Medical Association, 240(5), 563-569 (2012-02-16)
To compare the effects of 2 preoperative anti-inflammatory regimens on intraocular inflammation following phacoemulsification. Randomized controlled trial 21 dogs with immature cataracts. All dogs had cataract surgery via phacoemulsification, and most received prosthetic intraocular lenses. Dogs were randomly divided into
Susan M Wakil et al.
Canadian journal of ophthalmology. Journal canadien d'ophtalmologie, 47(4), 339-343 (2012-08-14)
The objective of this study was to present a unique case of unilateral facial nerve palsy as an isolated complication of herpes zoster ophthalmicus. An 82-year-old immunocompetent male presented with a 1-week history of painful left scalp lesions. The diagnosis
Florian Birnbaum et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 251(1), 95-103 (2012-05-11)
Postoperative astigmatism following penetrating keratoplasty is a major problem after corneal transplantation. The main goal of new trephination techniques such as femtosecond laser or excimer-laser trephination is to improve refractive and visual outcomes. The femtosecond laser technique makes profiled corneal
Jatin N Ashar et al.
The British journal of ophthalmology, 97(11), 1391-1394 (2013-03-29)
To evaluate a step ladder approach for immunosuppressive regimen for Mooren's ulcer. We retrospectively analysed patients of Mooren's ulcer presenting to a tertiary care centre in south India from 1987 to 2010. Patients were analysed for the age, disease severity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service