Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46632

Supelco

Citric acid sesquipiperazine salt hydrate

VETRANAL®, analytical standard

Synonym(s):

tri-Piperazine dicitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H10N2)3 · (C6H8O7)2 · xH2O
CAS Number:
Molecular Weight:
321.33 (anhydrous basis)
Beilstein/REAXYS Number:
3777748
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

OC(C(CC(O)=O)(CC(O)=O)O)=O.OC(C(CC(O)=O)(O)CC(O)=O)=O.O.N1CCNCC1.N2CCNCC2.N3CCNCC3

InChI

1S/2C6H8O7.3C4H10N2.H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;3*1-2-6-4-3-5-1;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);3*5-6H,1-4H2;1H2

InChI key

LWMBPKJYEQGDLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

Tyr-Gly-Gly-Phe-Leu-Arg-Arg-Gln-Phe-Lys-Val-Val-Thr.nCH3COOH

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Treatment of helminthic diseases.
J L Lefrock et al.
American family physician, 32(3), 182-188 (1985-09-01)
Samuel Ghasi et al.
American journal of therapeutics, 19(2), 81-87 (2010-03-11)
The effect of piperazine citrate on the electrocardiogram of the rat after treatment with piperazine citrate at 30, 60, and 100 mg/kg body weight for 16 weeks was investigated. The results were compared with a control group. There was prolongation
Piperazine toxicity in Afghan puppies.
R C Bownass
The Veterinary record, 120(13), 310-310 (1987-03-28)
P S Kamath et al.
Gastroenterology, 91(3), 730-732 (1986-09-01)
Roundworms migrating into the biliary tract are a relatively common cause of biliary symptoms in areas where the parasite is endemic. Visualization of worms by conventional radiologic methods is often unsatisfactory, whereas ultrasonography offers a rapid, safe, and noninvasive approach
D P Britt et al.
The Veterinary record, 123(9), 219-221 (1988-08-27)
The presence of benzimidazole-resistant strains of equine small strongyles was confirmed in horses at two properties in north west England by a series of faecal egg counts and larval cultures after treatment with mebendazole. A trail formulation of mebendazole in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service