46420
Megestrol acetate
VETRANAL®, analytical standard
Synonym(s):
17α-Acetoxy-6-methyl-4,6-pregnadiene-3,20-dione, 17α-Hydroxy-6-methyl-4,6-pregnadiene-3,20-dione 17-acetate, 17α-Hydroxy-6-methyl-6-dehydroprogesterone 17-acetate, Megestrol 17-acetate
About This Item
grade
analytical standard
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
[H][C@]12CC[C@@]3(C)[C@@]([H])(CC[C@]3(OC(C)=O)C(C)=O)[C@]1([H])C=C(C)C4=CC(=O)CC[C@]24C
InChI
1S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h12-13,18-20H,6-11H2,1-5H3/t18-,19+,20+,22-,23+,24+/m1/s1
InChI key
RQZAXGRLVPAYTJ-GQFGMJRRSA-N
Gene Information
human ... PGR(5241)
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service