Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46316

Supelco

Fluazinam solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Synonym(s):

3-Chloro-N-(3-chloro-2,6-dinitro-4-trifluoromethylphenyl)-5-trifluoromethyl-2-pyridylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H4Cl2F6N4O4
CAS Number:
Molecular Weight:
465.09
Beilstein/REAXYS Number:
4772672
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

−20°C

SMILES string

[O-][N+](=O)c1cc(c(Cl)c(c1Nc2ncc(cc2Cl)C(F)(F)F)[N+]([O-])=O)C(F)(F)F

InChI

1S/C13H4Cl2F6N4O4/c14-6-1-4(12(16,17)18)3-22-11(6)23-9-7(24(26)27)2-5(13(19,20)21)8(15)10(9)25(28)29/h1-3H,(H,22,23)

InChI key

UZCGKGPEKUCDTF-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epaminondas J Paplomatas et al.
Pest management science, 61(7), 691-698 (2005-03-02)
The molecular profile and the biological response of isolates of Pyricularia oryzae Cavara obtained from ctenanthe to two strobilurins (azoxystrobin, kresoxim-methyl) and the phenylpyridinamine fungicide fluazinam were characterized, and compared with isolates from rice plants. Five different isozymes (alpha-esterase, lactate
D P Bruynzeel et al.
Contact dermatitis, 33(1), 8-11 (1995-07-01)
We describe an outbreak of contact dermatitis in a tulip bulb processing company. Shortly after the introduction of a new pesticide, the fungicide fluazinam, employees started to complain of dermatitis of the arms and the face. 8 employees were investigated
Jeong Eun Lee et al.
Neurochemistry international, 60(8), 773-781 (2012-04-03)
Although the underlying cause of Parkinson's disease (PD) is not well characterized, epidemiological studies suggest that exposure to agricultural chemicals is a risk factor for PD. Fluazinam (FZN) is a new active ingredient for the control of grey mould, belonging
A Draper et al.
Occupational and environmental medicine, 60(1), 76-77 (2002-12-25)
We report two cases of occupational asthma caused by sensitisation to powdered fungicides fluazinam and chlorothalonil, from the same fungicide formulation plant. Both developed work related lower respiratory symptoms after a latent interval of asymptomatic exposure. The diagnosis in each
J D M Belgers et al.
Ecotoxicology and environmental safety, 72(2), 579-584 (2008-08-16)
We tested the sensitivity of nine submersed macrophyte species to the fungicides chlorothalonil, pentachlorophenol, fluazinam, and carbendazim. Endpoints determined 3 weeks after the start of the treatment were based on shoot and root growth in water. Carbendazim proved not or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service