Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46247

Supelco

Equilin

VETRANAL®, analytical standard

Synonym(s):

1,3,5(10),7-Estratetraen-3-ol-17-one, 3-Hydroxy-1,3,5(10),7-estratetraen-17-one, 7-Dehydroestrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20O2
CAS Number:
Molecular Weight:
268.35
Beilstein/REAXYS Number:
2624302
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

238-240 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

C[C@]12CC[C@H]3C(=CCc4cc(O)ccc34)[C@@H]1CCC2=O

InChI

1S/C18H20O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3-5,10,14,16,19H,2,6-9H2,1H3/t14-,16+,18+/m1/s1

InChI key

WKRLQDKEXYKHJB-HFTRVMKXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazard

signalword

Warning

Hazard Classifications

Carc. 2 - Lact. - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ekaterina V Rokhina et al.
The Science of the total environment, 417-418, 280-290 (2012-01-17)
Estrone (E1), 17β-estradiol (E2), estriol (E3), equilin (EQ) and 17α-estradiol (17α) estrogen hormones are released by humans and animals and have been detected in the environment and municipal wastewater treatment plants. The structural and electronic properties of natural hormone molecules
Rupinder K Bhamra et al.
Menopause (New York, N.Y.), 18(4), 393-399 (2010-11-26)
A randomized, parallel-design study was conducted to determine the pharmacokinetic profile of synthetic conjugated estrogens A (SCE-A) vaginal cream (0.625 mg SCE-A/g) when administered at intervals (1 g once daily for 7 d, then twice weekly) over a 27-day period
Maher A Abdallah et al.
Fertility and sterility, 87(4), 978-981 (2007-01-09)
A combination of E and progestogens significantly increased matrix metalloproteinase (MMP)-2 in both T47D cells (E(2)-medroxyprogesteroneacetate [MPA] and E(2)-P) and MCF-7 cells (E(2)-MPA, E(2)-P, and equilin-MPA). All combinations resulted in higher MMP-9 levels in MCF-7 cells, but higher MMP-9 levels
John C Reepmeyer et al.
Journal of chromatography. A, 1083(1-2), 42-51 (2005-08-05)
Equilin-3-sulfate and delta8,9-dehydroestrone-3-sulfate are two isomers found in equine conjugated estrogens that differ in structure only by the position of a double bond in the steroid B-ring. These geometric isomers were not resolved on a C18 column during the analysis
Dieldrich S Bermudez et al.
International journal of andrology, 35(3), 397-406 (2012-05-23)
There is growing concern of exposure of fish, wildlife and humans to water sources contaminated with oestrogens and the potential impact on reproductive health. Environmental oestrogens can come from various sources including concentrated animal feedlot operations (CAFO), municipal waste, agricultural

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service