Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46159

Supelco

Monocrotophos solution

10 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14NO5P
CAS Number:
Molecular Weight:
223.16
Beilstein/REAXYS Number:
8415832
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

CNC(=O)\C=C(/C)OP(=O)(OC)OC

InChI

1S/C7H14NO5P/c1-6(5-7(9)8-2)13-14(10,11-3)12-4/h5H,1-4H3,(H,8,9)/b6-5+

InChI key

KRTSDMXIXPKRQR-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F

flash_point_c

2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amajad Iqbal Kazi et al.
Neurotoxicology, 33(2), 156-161 (2012-01-31)
Neurotoxicity of organophosphate pesticide poisoning, a lead cause of death in South Asia, has not been clearly elucidated. Organophosphates inhibit acetylcholinesterase and neurotoxicity is primarily a result of acetylcholine induced hyperactivation in different regions of the brain. Neurotoxicity also results
Madhu Lata Sankhwar et al.
Human & experimental toxicology, 31(6), 606-616 (2011-04-22)
Studies on the neurobehavioral toxicity of monocrotophos, an organophosphate, have been carried out on rats following their exposure from postnatal day (PD) 22 to PD 49 to investigate whether neurobehavioral changes are transient or persistent. Exposure of rats to monocrotophos
Madhusudan Reddy Narra et al.
Toxicology and industrial health, 28(4), 343-352 (2011-10-11)
Sublethal effects of chlorpyrifos (CPF) and monocrotophos (MCP) on fish biochemical constituents were investigated along with the assessment of recovery response after cessation of intoxication. The fish, Clarias batrachus were exposed to 1.656 mg(-l) and 2.114 mg(-l) of CPF and
Deok Ho Kwon et al.
Insect biochemistry and molecular biology, 42(3), 212-219 (2011-12-27)
The mutations (G228S, A391T and F439W) and duplication of the acetylcholinesterase (AChE) gene (Tuace) are involved in monocrotophos resistance in the two-spotted spider mites, Tetranychus urticae (Kwon et al., 2010a, b). The overexpression of T. urticae AChE (TuAChE) as a result of
Hua Tian et al.
Toxicology and applied pharmacology, 263(2), 163-170 (2012-06-21)
Monocrotophos is a highly toxic organophosphorus pesticide that has been confirmed to be an endocrine-disrupting chemical. To evaluate the influence of this pollutant on the reproductive system of male fish, we studied the sex steroid levels, reproductive traits, sex ratio

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service