46141
Ethidium bromide/SYBR® green replacement cartridges
binding capacity: 50 mg
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.51
Recommended Products
Application
5 replacement cartridges for decontamination of solutions containing Ethidium bromide or SYBR® Green; binding capacity for above mentioned fluorescent stains: 50 mg
Legal Information
SYBR is a registered trademark of Life Technologies
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service