Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46128

Sigma-Aldrich

Eucalyptol

Ph. Helv.

Synonym(s):

Cineolum, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein/REAXYS Number:
105109
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

agency

Ph. Helv.
USP/NF

refractive index

n20/D 1.457 (lit.)

bp

176-177 °C (lit.)

mp

1-2 °C (lit.)

density

0.921 g/mL at 25 °C (lit.)

SMILES string

C[C@]12CC[C@H](CC1)C(C)(C)O2

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3/t8-,10+

InChI key

WEEGYLXZBRQIMU-WAAGHKOSSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

125.6 °F - closed cup

flash_point_c

52 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee-jin Jun et al.
Bioorganic & medicinal chemistry letters, 23(2), 579-583 (2012-12-19)
We investigated the effect of cineole on the expression of genes related to reverse cholesterol transport and hepatic fatty acid metabolism. Cineole, a small aroma compound in teas and herbs, significantly stimulated the transactivation of liver X receptor modulator (LXR)-α
Zerihun A Demissie et al.
Plant molecular biology, 79(4-5), 393-411 (2012-05-18)
Several members of the genus Lavandula produce valuable essential oils (EOs) that are primarily constituted of the low molecular weight isoprenoids, particularly monoterpenes. We isolated over 8,000 ESTs from the glandular trichomes of L. x intermedia flowers (where bulk of
Lisa A Shipley et al.
Journal of chemical ecology, 38(9), 1178-1189 (2012-10-12)
Pygmy rabbits (Brachylagus idahoensis) are one of only three vertebrates that subsist virtually exclusively on sagebrush (Artemisia spp.), which contains high levels of monoterpenes that can be toxic. We examined the mechanisms used by specialist pygmy rabbits to eliminate 1,8-cineole
Johannes F-W Greiner et al.
Biochimica et biophysica acta, 1833(12), 2866-2878 (2013-07-23)
Natural plant-derived products are commonly applied to treat a broad range of human diseases, including cancer as well as chronic and acute airway inflammation. In this regard, the monoterpene oxide 1,8-cineol, the active ingredient of the clinically approved drug Soledum®
Anke Fähnrich et al.
Plant molecular biology, 85(1-2), 135-145 (2014-02-05)
Nicotiana species of the section Alatae emit a characteristic floral scent comprising the' cineole cassette' monoterpenes 1,8-cineole, limonene, myrcene, β-pinene, α-pinene, sabinene and α-terpineol. All previously isolated 'cineole cassette'-monoterpene synthase genes are multi product enzymes that synthesize the seven compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service