Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

45979

Supelco

o-Toluidine

analytical standard

Synonym(s):

2-Aminotoluene, 2-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein/REAXYS Number:
741981
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.7 (vs air)

vapor pressure

0.26 mmHg ( 25 °C)

autoignition temp.

899 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.572 (lit.)

bp

199-200 °C (lit.)
89-90 °C/11 mmHg (lit.)

mp

−28 °C (lit.)

solubility

H2O: slightly soluble

density

1.008 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cc1ccccc1N

InChI

1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3

InChI key

RNVCVTLRINQCPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Toluidine is one of the metabolites of prilocaine and a causative agent of methaemoglobinemia, associated with exceeding doses of prilocaine.[1]

Application

o-Toluidine may be used as a reference standard for the determination of o-toluidine in water samples by gas chromatographic method following solid-phase extraction and derivatization with iodine.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatographic determination of aromatic amines in water samples after solid-phase extraction and derivatization with iodine: I. Derivatization.
Schmidt TC, et al.
Journal of Chromatography A, 810(1-2), 161-172 (1998)
Simultaneous determination of lidocaine, prilocaine and the prilocaine metabolite o-toluidine in plasma by high-performance liquid chromatography.
Klein J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 655(1), 83-88 (1994)
Hun-Je Jo et al.
Chemosphere, 70(6), 949-957 (2007-09-26)
In this study, major toxicants in pigment wastewaters (i.e. raw wastewater and effluent) were determined by toxicity identification evaluations (TIEs), and changes in their toxicities due to combined gamma-ray (gamma-ray) and ozone treatments were evaluated. From TIE results using Daphnia
Tania Carreón et al.
Occupational and environmental medicine, 67(5), 348-350 (2009-11-04)
In 1991, the US National Institute for Occupational Safety and Health (NIOSH) reported an increased bladder cancer risk in a cohort of 1749 workers potentially exposed to o-toluidine and aniline at a chemical manufacturing plant. As additional information showed that
Ivo Kalkman et al.
Physical chemistry chemical physics : PCCP, 11(21), 4311-4318 (2009-05-22)
The rotationally resolved spectrum of the o-toluidine S(1)<-- S(0) origin was measured using laser induced fluorescence spectroscopy. From the resulting spectrum torsional barriers to internal rotation of the methyl group were derived, which resulted in S(0) state values of V(3)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service