Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45969

Supelco

2,4-Dinitrotoluene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)2
CAS Number:
Molecular Weight:
182.13
Beilstein/REAXYS Number:
1912834
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 102.7 °C)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

67-70 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1

InChI

1S/C7H6N2O4/c1-5-2-3-6(8(10)11)4-7(5)9(12)13/h2-4H,1H3

InChI key

RMBFBMJGBANMMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dinitrotoluene is classified under the group of nitroaromatic compounds, which are organic nitro derivatives of analytical research interest. It finds wide use as a precursor of 2,4,6-trinitrotoluene (TNT) explosive as well as dyes and urethane products. 2,4-Dinitrotoluene is regarded as a potential threat to human health even when present in trace amounts.[1][2]

Application

2,4-Dinitrotoluene may be used as an analytical standard for the determination of the analyte in forensic samples, explosive-contaminated groundwater samples, industrial effluents and seawater samples by various chromatographic techniques.[3][1][4][2][5][6]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

311.0 °F - closed cup

flash_point_c

155.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coupled solid phase extraction-supercritical fluid extraction-on-line gas chromatography of explosives from water
Slack GC, et al.
Journal of High Resolution Chromatography, 16(8), 473-478 (1993)
Determination of dinitrotoluene isomers in sea water and industrial effluent by high-resolution electron-capture gas chromatography with a glass capillary column
Hashimoto A, et al.
Analyst, 105(1253), 787-793 (1980)
E Psillakis et al.
Journal of chromatography. A, 907(1-2), 211-219 (2001-02-24)
The application of solvent microextraction to the analysis of nitroaromatic explosives is presented. Extraction of 11 nitroaromatics was achieved by suspending 1 microl of organic solvent to the tip of a microsyringe in a stirred aqueous solution. Parameters such as
Application of high-field proton nuclear magnetic resonance (1 H-NMR) spectroscopy for the analysis of explosives and related compounds in groundwater samples-a comparison with the high-performance liquid chromatography (HPLC) method
Preiss A, et al.
Fresenius Journal of Analytical Chemistry, 356(7), 445-451 (1996)
Jingbo Xu et al.
Journal of hazardous materials, 203-204, 299-307 (2012-01-03)
In this study relative condition factor (RCF) and hepatosomatic index (HSI) as well as the available energy reserves of common carp (Cyprinus carpio) by 2,4-DNT semi-static bioassay were determined and linked to effects of enzymes in liver tissues. Fish were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service