Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45948

Supelco

2,3-Dichlorotoluene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3Cl2
CAS Number:
Molecular Weight:
161.03
Beilstein/REAXYS Number:
2412384
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.551 (lit.)

bp

207-208 °C (lit.)

mp

6 °C (lit.)

density

1.228 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1cccc(Cl)c1Cl

InChI

1S/C7H6Cl2/c1-5-3-2-4-6(8)7(5)9/h2-4H,1H3

InChI key

GWLKCPXYBLCEKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

181.4 °F - closed cup

flash_point_c

83 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riquan Liao et al.
Environmental monitoring and assessment, 191(5), 309-309 (2019-04-28)
To enhance our understanding on environmental conditions of the Maowei Sea in Guangxi province, China, the concentration and distribution of 22 chlorobenzene compounds (CBs) in the surface sediment were determined by gas chromatography-mass spectrometry (GC-MS). The relationship of the sediment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service