45861
Heptachlor exo-epoxide solution
100 μg/mL in methanol, PESTANAL®, analytical standard, isomer B
About This Item
grade
analytical standard
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
concentration
100 μg/mL in methanol
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care
format
single component solution
storage temp.
2-8°C
SMILES string
[H][C@]12O[C@@]1([H])[C@@]3([H])[C@@]([H])([C@H]2Cl)C4(Cl)C(Cl)=C(Cl)C3(Cl)C4(Cl)Cl
InChI
1S/C10H5Cl7O/c11-3-1-2(4-5(3)18-4)9(15)7(13)6(12)8(1,14)10(9,16)17/h1-5H/t1-,2+,3+,4-,5+,8?,9?/m0/s1
InChI key
ZXFXBSWRVIQKOD-TTWWGNBVSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
51.8 °F - closed cup
flash_point_c
11 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service