Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45852

Supelco

Endosulfan solution

100 μg/mL in hexane, PESTANAL®, analytical standard, mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6Cl6O3S
CAS Number:
Molecular Weight:
406.93
Beilstein/REAXYS Number:
2062338
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

ClC1=C(Cl)C2(Cl)C3COS(=O)OCC3C1(Cl)C2(Cl)Cl

InChI

1S/C9H6Cl6O3S/c10-5-6(11)8(13)4-2-18-19(16)17-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3?,4?,7-,8+,19?

InChI key

RDYMFSUJUZBWLH-QTSFFCDASA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-14.8 °F - closed cup

flash_point_c

-26 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jimmy Yuk et al.
Environmental pollution (Barking, Essex : 1987), 175, 35-44 (2013-01-22)
One-dimensional (1-D) and two-dimensional (2-D) nuclear magnetic resonance (NMR)-based metabolomics was used to investigate the toxic mode of action (MOA) of endosulfan, an organochlorine pesticide, and its degradation product, endosulfan sulfate, to Eisenia fetida earthworms in soil. Three soil concentrations
Carlos Leandro Negro et al.
Bulletin of environmental contamination and toxicology, 89(5), 1000-1003 (2012-09-11)
We examined the bioaccumulation of α- and β-endosulfan and endosulfan sulfate in tissues from the crab Zilchiopsis collastinensis. There was more endosulfan accumulated in the hepatopancreas (from <2 to 467.8 ng g(-1)) than in the gonads (from <2 to 52.1
Ryota Kataoka et al.
Applied microbiology and biotechnology, 97(8), 3285-3292 (2013-03-07)
Endosulfan and endosulfan sulfate are persistent organic pollutants that cause serious environmental problems. Although these compounds are already prohibited in many countries, residues can be detected in soils with a history of endosulfan application. Endosulfan is transformed in the environment
Acute kidney injury in endosulfan poisoning.
Manjusha Yadla et al.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia, 24(3), 592-593 (2013-05-04)
C L Negro et al.
Ecotoxicology and environmental safety, 90, 121-127 (2013-01-29)
Biocides are periodically applied in agricultural activities, reaching aquatic systems and acting upon the biota. Amphipods are widely used in toxicity tests because of their sensitivity to a wide range of pollutants. In this work, we report the differential lethality

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service