Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45828

Supelco

Chlorfenvinphos solution

100 μg/mL in cyclohexane, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14Cl3O4P
CAS Number:
Molecular Weight:
359.57
Beilstein/REAXYS Number:
8712900
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

description

mixture of cis and trans isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in cyclohexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

CCOP(=O)(OCC)O\C(=C/Cl)c1ccc(Cl)cc1Cl

InChI

1S/C12H14Cl3O4P/c1-3-17-20(16,18-4-2)19-12(8-13)10-6-5-9(14)7-11(10)15/h5-8H,3-4H2,1-2H3/b12-8-

InChI key

FSAVDKDHPDSCTO-WQLSENKSSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

-0.4 °F - closed cup

flash_point_c

-18 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

María A Martínez et al.
Journal of analytical toxicology, 36(1), 44-51 (2012-02-01)
Chlorfenvinphos (CFVP) is an organophosporus insecticide designated as a threat agent by the National Institutes of Health (NIH). However, there are very few reported cases of poisonings in humans and none with postmortem toxicological analysis. We report the first two
Naotaka Kakoi et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 129(10), 1255-1264 (2009-10-03)
Heparin is widely used as an anticoagulant for the treatment and prevention of thrombotic disorders. Recently, hundreds of cases of anaphylactic reaction as adverse effects were reported by the presence of contaminating oversulfated chondroitin sulfate (OSCS) in some heparin preparations.
Grzegorz Raszewski et al.
Basic & clinical pharmacology & toxicology, 105(1), 37-45 (2009-04-18)
The ability of obidoxime with atropine and diazepam mixture to reactivate acetylcholinesterase inhibited by the organophosphorus compound chlorfenvinphos was compared in the central nervous system and peripheral tissues of rats. The animals were intoxicated with chlorfenvinphos (6 mg/kg, p.o.) and
Georges Istamboulie et al.
Talanta, 77(5), 1627-1631 (2009-01-23)
This work shows the possibility of combining the high sensitivity of genetically-modified Drosophila melanogaster acetylcholinesterase (B394) with the ability of phosphotriesterase (PTE) to hydrolyse organophosphate compounds, in the aim of developing a biosensor selective to two insecticides of interest: chlorpyrifos
Colin J Jackson et al.
Applied and environmental microbiology, 75(15), 5153-5156 (2009-06-09)
In silico substrate docking of both stereoisomers of the pesticide chlorfenvinphos (CVP) in the phosphotriesterase from Agrobacterium radiobacter identified two residues (F131 and W132) that prevent productive substrate binding and cause stereospecificity. A variant (W131H/F132A) was designed that exhibited ca.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service