Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45682

Supelco

1,2-Dichloro-3-nitrobenzene

PESTANAL®, analytical standard

Synonym(s):

vic.-Nitro-o-dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NO2
CAS Number:
Molecular Weight:
192.00
Beilstein/REAXYS Number:
2048029
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

257-258 °C (lit.)

mp

57-61 °C (lit.)

density

1.449 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

[O-][N+](=O)c1cccc(Cl)c1Cl

InChI

1S/C6H3Cl2NO2/c7-4-2-1-3-5(6(4)8)9(10)11/h1-3H

InChI key

CMVQZRLQEOAYSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

305.6 °F

flash_point_c

152 °C

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darcy Fallaise et al.
Magnetic resonance in chemistry : MRC (2018-12-18)
Nonaqueous phase liquids (NAPLs) located at the surface of the water table and/or below the water table are often a significant source for groundwater contamination near current or former commercial/industrial facilities. Due to the complex and long history of many

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service