Sign In to View Organizational & Contract Pricing.
About This Item
Linear Formula:
HOCH2[CH(OH)]2CH2OH
CAS Number:
Molecular Weight:
122.12
EC Number:
205-737-3
UNSPSC Code:
12352201
MDL number:
Beilstein/REAXYS Number:
1719753
Skip To
assay
≥99.0% (HPLC)
bp
329-331 °C (lit.)
mp
118-120 °C (lit.), 118-121 °C
solubility
H2O: 0.1 g/mL, clear to almost clear, colorless
SMILES string
OC[C@@H](O)[C@@H](O)CO
InChI
1S/C4H10O4/c5-1-3(7)4(8)2-6/h3-8H,1-2H2/t3-,4+
InChI key
UNXHWFMMPAWVPI-ZXZARUISSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Allelic variation of the Tas1r3 gene affects behavioral taste responses to this sugar alcohol, suggesting that it is a T1R3 receptor ligand.
comparable product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service