Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45658

Supelco

Secbumeton

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N5O
CAS Number:
Molecular Weight:
225.29
Beilstein/REAXYS Number:
8981797
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(NC(C)CC)nc(OC)n1

InChI

1S/C10H19N5O/c1-5-7(3)12-9-13-8(11-6-2)14-10(15-9)16-4/h7H,5-6H2,1-4H3,(H2,11,12,13,14,15)

InChI key

ZJMZZNVGNSWOOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Secbumeton belongs to the class of triazine herbicides, widely used in agriculture to obtain high yield productions. Its mode of action involves the inhibition of photosynthesis, thereby controlling the growth of grasses and broadleaf weeds.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Secbumeton may be used as an analytical reference standard for the quantification of the analyte in environmental samples, agricultural commodities and vegetable samples using different chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersive liquid-liquid microextraction method based on solidification of floating organic droplet for the determination of triazine herbicides in water and sugarcane samples
Sanagi MM, et al.
Food Chemistry, 133(2), 557-562 (2012)
[Histological, histochemical and histoenzymatic changes at the level of the rat liver under the action of secbumetone].
T Gheorghiu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 90(4), 693-695 (1986-10-01)
Determination of triazine herbicides in vegetables by ionic liquid foam floatation solid phase extraction high performance liquid chromatography
Zhang L, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 953(2), 132-137 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service