Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45647

Supelco

Pyranocoumarin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18O4
CAS Number:
Molecular Weight:
322.35
Beilstein/REAXYS Number:
290680
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

COC1(C)CC(c2ccccc2)C3=C(O1)c4ccccc4OC3=O

InChI

1S/C20H18O4/c1-20(22-2)12-15(13-8-4-3-5-9-13)17-18(24-20)14-10-6-7-11-16(14)23-19(17)21/h3-11,15H,12H2,1-2H3

InChI key

ZGFASEKBKWVCGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pyranocoumarin may be used as an analytical reference standard for the quantification of the analyte in Ammi visnaga L. fruits and pharmaceuticals using reversed-phase high-performance liquid chromatography (RP-HPLC).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M He et al.
Archives of biochemistry and biophysics, 372(1), 16-28 (1999-11-24)
Possible reasons for the observed differences in metabolic behavior and drug interaction liability between the structurally similar oral anticoagulants warfarin and phenprocoumon were explored. Incubating (S)-phenprocoumon with human liver microsomes and cDNA-expressed CYP2C9 and determining its metabolism both in the
A S Bourinbaiar et al.
Acta virologica, 37(4), 241-250 (1993-08-01)
A single dose of coumarin derivatives, warfarin, 4-hydroxycoumarin and umbelliferone, added at the time of inoculation either by free virus or by contact with U1 monocytes exhibited a dose-dependent inhibitory effect on viral replication in target MOLT-4 lymphocytes observable even
Junsei Taira et al.
Antioxidants (Basel, Switzerland), 8(12) (2019-12-11)
This study focused on exploring the nuclear factor-erythroid-2-related factor (Nrf2) active compound to avoid oxidative stress related to various diseases, such as obesity and diabetes mellitus. The activity of the Nrf2-ARE (antioxidant response element) signaling was evaluated by a reporter
L D Heimark et al.
Journal of medicinal chemistry, 27(8), 1092-1095 (1984-08-01)
An octanol/water model system and circular dichroism (CD) spectroscopy have been used to study the solution conformation of warfarin in aqueous and lipid environments. Upon partitioning of (S)-warfarin from buffer pH 7.4 into octanol, the position of the absorption band
G Zgórka et al.
Journal of chromatography. A, 797(1-2), 305-309 (1998-05-16)
A new, simple and rapid solid-phase extraction method for the determination of furanochromones and pyranocoumarins in Ammi visnaga L. fruits and pharmaceuticals by reversed-phase high-performance liquid chromatography (RP-HPLC) was developed. The isolation of compounds examined was carried out on octadecyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service