Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

456446

SAFC

Decahydronaphthalene, mixture of cis + trans

97%

Synonym(s):

Decalin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein/REAXYS Number:
878165
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

vapor density

4.76 (vs air)

Quality Level

vapor pressure

42 mmHg ( 92 °C)
741 mmHg ( 188 °C)

assay

97%

autoignition temp.

482 °F

expl. lim.

0.7-4.9 %, 100 °F

refractive index

n20/D 1.474 (lit.)

bp

189-191 °C (lit.)
189-191 °C

mp

−125 °C (lit.)

density

0.896 g/mL at 25 °C (lit.)

SMILES string

C1CCC2CCCCC2C1

InChI

1S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10H,1-8H2

InChI key

NNBZCPXTIHJBJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Decalin is a trademark of Sigma-Aldrich Co. LLC

recommended

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

134.6 °F - closed cup

flash_point_c

57 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conformational isomerization of cis-decahydronaphthalene over zeolite catalysts.
Lai WC and Song C.
Catalysis Today, 31(1), 171-181 (1996)
The Densities and Surface Tensions of cis-and trans-Decahydronaphthalene between-30 and 180?1.
Seyer WF and Davenport CH.
Journal of the American Chemical Society, 63(9), 2425-2427 (1941)
Poly (diketopyrrolopyrrole-benzothiadiazole) with Ambipolarity Approaching 100% Equivalency.
Cho S, et al.
Advances in Functional Materials, 21(10), 1910-1916 (2011)
Kinetics of liquid phase cyclohexene hydrogenation on Pd-Al/biomorphic carbon catalysts.
Caza?a F, et al.
Catalysis Today, 13, 14-14 (2014)
Ken K Qian et al.
Journal of pharmaceutical sciences, 100(7), 2801-2815 (2011-02-22)
Spontaneous crystalline-to-amorphous phase transformation of organic or medicinal molecules in the presence of mesoporous materials has been observed, for which pathway was suggested to be via the vapor phase, that is, sublimation of the crystalline molecules followed by adsorption on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service